Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prl2b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138861.1 | NP_620216.1 | 4 - 5 | 551 | 99 | |
GenBank mRNA | AB022882.1 | - | 4 - 5 | 548 | 99 | |
AF234635.1 | - | 4 - 5 | 551 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 3' Most Rattus norvegicus 1700 Probe spans exons |