Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P4ha1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172062.2 | NP_742059.2 | 2 - 3 | 313 | 93 | |
XM_006256424.2 | XP_006256486.1 | 3 - 4 | 398 | 93 | ||
XM_006256425.2 | XP_006256487.1 | 2 - 3 | 307 | 93 | ||
GenBank mRNA | BC078703.1 | - | 2 - 3 | 313 | 7115543 | 93 |
X78949.1 | - | 2 - 3 | 147 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |