Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppil3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175707.3 | NP_783638.1 | 1 - 2 | 267 | 106 | |
GenBank mRNA | AF315802.1 | - | 1 - 2 | 55 | 106 | |
BC087645.1 | - | 1 - 2 | 267 | 7311008 | 106 | |
FQ224392.1 | - | 1 - 2 | 64 | 106 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |