Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cct3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199091.2 | NP_954522.1 | 1 - 2 | 43 | 63 | |
GenBank mRNA | BC063178.1 | - | 1 - 2 | 47 | 6890550 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |