Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Brca1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012514.1 | NP_036646.1 | 15 - 16 | 4935 | 87 | |
XM_008768092.1 | XP_008766314.1 | 15 - 16 | 5151 | 87 | ||
GenBank mRNA | AF036760.1 | - | 15 - 16 | 4935 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |