Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acadvl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012891.2 | NP_037023.1 | 13 - 14 | 1354 | 139 | |
GenBank mRNA | BC087036.1 | - | 13 - 14 | 1375 | 7193535 | 139 |
D30647.1 | - | 13 - 14 | 1356 | 139 | ||
FQ219329.1 | - | 13 - 14 | 1399 | 139 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |