Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ntf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013184.3 | NP_037316.2 | 1 - 2 | 79 | 128 | |
GenBank mRNA | S69323.1 | - | 1 - 2 | 79 | 128 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Inventoried Probe spans exons |