Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gabra6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021841.1 | NP_068613.1 | 4 - 5 | 447 | 106 | |
XM_006246128.2 | XP_006246190.1 | 4 - 5 | 596 | 106 | ||
XM_008767636.1 | XP_008765858.1 | 2 - 3 | 518 | 106 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Ion Channel Detect Genomic DNA Rattus norvegicus Probe spans exons |