Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pnpo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022601.2 | NP_072123.1 | 6 - 7 | 747 | 103 | |
GenBank mRNA | BC087016.1 | - | 6 - 7 | 747 | 7128621 | 103 |
U91561.1 | - | 6 - 7 | 738 | 103 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |