Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sv2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031593.1 | NP_113781.1 | 5 - 6 | 1272 | 62 | |
GenBank mRNA | AF060174.1 | - | 5 - 6 | 1272 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |