Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdk4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053551.1 | NP_446003.1 | 5 - 6 | 721 | 64 | |
GenBank mRNA | AF034577.1 | - | 5 - 6 | 721 | 64 | |
FQ216954.1 | - | 764 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Detect Genomic DNA Rattus norvegicus Probe spans exons |