Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bdh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053995.3 | NP_446447.2 | 3 - 4 | 360 | 74 | |
XM_006248478.2 | XP_006248540.1 | 3 - 4 | 416 | 74 | ||
XM_008768691.1 | XP_008766913.1 | 2 - 3 | 198 | 74 | ||
GenBank mRNA | BC085916.1 | - | 3 - 4 | 360 | 7108483 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |