Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ripk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139342.1 | NP_647558.1 | 6 - 7 | 1078 | 95 | |
XM_006251928.2 | XP_006251990.1 | 5 - 6 | 1930 | 95 | ||
XM_008770661.1 | XP_008768883.1 | 4 - 5 | 891 | 95 | ||
XM_008770662.1 | XP_008768884.1 | 5 - 6 | 1262 | 95 | ||
GenBank mRNA | AF036537.2 | - | 6 - 7 | 1078 | 95 | |
BC158580.1 | - | 4 - 5 | 645 | 7378607 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |