Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpinh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017173.1 | NP_058869.1 | 3 - 4 | 838 | 63 | |
GenBank mRNA | BC086529.1 | - | 2 - 3 | 729 | 7303866 | 63 |
M69246.1 | - | 3 - 4 | 838 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |