Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acsm4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181695.1 | NP_859046.1 | 8 - 9 | 1247 | 78 | |
GenBank mRNA | AB096688.1 | - | 8 - 9 | 1247 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |