Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Micalcl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_182669.2 | NP_872610.2 | 8 - 9 | 1944 | 103 | |
GenBank mRNA | AY149342.1 | - | 5 - 6 | 1384 | 103 | |
BC098647.1 | - | 8 - 9 | 1944 | 7112444 | 103 | |
BC100137.1 | - | 5 - 6 | 1398 | 7457561 | 103 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |