Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCOLCE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127640.1 | NP_001121112.1 | 1 - 2 | 597 | 73 | |
GenBank mRNA | BC161891.1 | - | 1 - 2 | 597 | 7189256 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |