Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mapkapk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012127.1 | NP_001012127.1 | 8 - 9 | 1121 | 71 | |
XM_006243776.2 | XP_006243838.1 | 8 - 9 | 1211 | 71 | ||
XM_008766531.1 | XP_008764753.1 | 8 - 9 | 1220 | 71 | ||
GenBank mRNA | BC081974.1 | - | 8 - 9 | 1121 | 7126359 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Rattus norvegicus Probe spans exons |