Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fastkd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191738.1 | NP_001178667.1 | 11 - 12 | 3152 | 108 | |
XM_006234311.1 | XP_006234373.1 | 11 - 12 | 1961 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |