Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106499.2 | NP_001099969.1 | 3 - 4 | 585 | 73 | |
GenBank mRNA | BC169083.1 | - | 3 - 4 | 585 | 7936869 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |