Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tspan18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107750.1 | NP_001101220.1 | 3 - 4 | 492 | 75 | |
XM_006234573.2 | XP_006234635.1 | 3 - 4 | 423 | 75 | ||
XM_006234574.2 | XP_006234636.1 | 4 - 5 | 565 | 75 | ||
XM_006234575.2 | XP_006234637.1 | 2 - 3 | 343 | 75 | ||
XM_008762027.1 | XP_008760249.1 | 2 - 3 | 243 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |