Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Amn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108061.1 | NP_001101531.1 | 8 - 9 | 912 | 101 | |
XM_006240588.2 | XP_006240650.1 | 8 - 9 | 895 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |