Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ak7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108055.3 | NP_001101525.2 | 7 - 8 | 910 | 66 | |
XM_008764805.1 | XP_008763027.1 | 7 - 8 | 1340 | 66 | ||
XM_008764806.1 | XP_008763028.1 | 6 - 7 | 1259 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Probe spans exons |