Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ak7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108055.3 | NP_001101525.2 | 8 - 9 | 1001 | 128 | |
XM_008764805.1 | XP_008763027.1 | 8 - 9 | 1431 | 128 | ||
XM_008764806.1 | XP_008763028.1 | 7 - 8 | 1350 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |