Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ak7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108055.3 | NP_001101525.2 | 9 - 10 | 1081 | 122 | |
XM_008764805.1 | XP_008763027.1 | 9 - 10 | 1511 | 122 | ||
XM_008764806.1 | XP_008763028.1 | 8 - 9 | 1430 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |