Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stra8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006236282.2 | XP_006236344.1 | 2 - 3 | 2638 | 85 | |
XM_575429.4 | XP_575429.2 | 1 - 2 | 381 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |