Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stra8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006236282.2 | XP_006236344.1 | 8 - 9 | 3475 | 70 | |
XM_575429.4 | XP_575429.2 | 7 - 8 | 1218 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Rattus norvegicus 1700 Probe spans exons |