Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asb15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006236158.2 | XP_006236220.1 | 9 - 10 | 1943 | 68 | |
XM_008762831.1 | XP_008761053.1 | 9 - 10 | 2099 | 68 | ||
XM_008762832.1 | XP_008761054.1 | 9 - 10 | 1900 | 68 | ||
XM_008762833.1 | XP_008761055.1 | 9 - 10 | 2121 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Rattus norvegicus 1700 Probe spans exons |