Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Luc7l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107853.1 | NP_001101323.1 | 5 - 6 | 847 | 88 | |
XM_006236314.2 | XP_006236376.1 | 7 - 8 | 686 | 88 | ||
XM_006236315.2 | XP_006236377.1 | 6 - 7 | 1350 | 88 | ||
XM_006236316.2 | XP_006236378.1 | 6 - 7 | 579 | 88 | ||
XM_006236319.2 | XP_006236381.1 | 5 - 6 | 1279 | 88 | ||
XM_006236321.2 | XP_006236383.1 | 5 - 6 | 917 | 88 | ||
XM_006236322.2 | XP_006236384.1 | 5 - 6 | 532 | 88 | ||
XM_006236323.2 | XP_006236385.1 | 7 - 8 | 1048 | 88 | ||
XM_008762778.1 | XP_008761000.1 | 6 - 7 | 3486 | 88 | ||
XM_008762779.1 | XP_008761001.1 | 6 - 7 | 3485 | 88 | ||
XM_008762780.1 | XP_008761002.1 | 6 - 7 | 786 | 88 | ||
XM_008762781.1 | XP_008761003.1 | 5 - 6 | 1299 | 88 | ||
GenBank mRNA | BC166855.1 | - | 5 - 6 | 868 | 9099059 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |