Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc9b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001173973.1 | NP_001167444.1 | 10 - 11 | 1613 | 114 | |
XM_008761518.1 | XP_008759740.1 | 9 - 10 | 1129 | 114 | ||
GenBank mRNA | BC158723.1 | - | 5 - 6 | 610 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |