Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Slc39a8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011952.1 | NP_001011952.1 | 4 - 5 | 930 | 108 | |
XM_006233356.2 | XP_006233418.1 | 5 - 6 | 969 | 108 | ||
XM_006233357.2 | XP_006233419.1 | 5 - 6 | 1189 | 108 | ||
GenBank mRNA | BC089844.1 | - | 4 - 5 | 930 | 7368889 | 108 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Rattus norvegicus Probe spans exons |