Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gstcd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107725.1 | NP_001101195.1 | 11 - 12 | 1998 | 123 | |
XM_008761505.1 | XP_008759727.1 | 10 - 11 | 3481 | 123 | ||
GenBank mRNA | FQ211741.1 | - | 10 - 11 | 1952 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |