Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bmpr1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024259.1 | NP_001019430.1 | 2 - 3 | 135 | 110 | |
XM_006233400.2 | XP_006233462.1 | 6 - 7 | 979 | 110 | ||
XM_006233401.2 | XP_006233463.1 | 5 - 6 | 640 | 110 | ||
XM_008761513.1 | XP_008759735.1 | 5 - 6 | 839 | 110 | ||
XM_008761514.1 | XP_008759736.1 | 8 - 9 | 1361 | 110 | ||
GenBank mRNA | BC092609.1 | - | 2 - 3 | 135 | 7310757 | 110 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Rattus norvegicus Probe spans exons |