Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkacb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077645.1 | NP_001071113.1 | 5 - 6 | 478 | 148 | |
XM_006233451.2 | XP_006233513.1 | 5 - 6 | 1195 | 148 | ||
XM_006233452.2 | XP_006233514.1 | 5 - 6 | 2700 | 148 | ||
XM_006233453.2 | XP_006233515.1 | 5 - 6 | 942 | 148 | ||
GenBank mRNA | D10770.1 | - | 5 - 6 | 478 | 148 |
Set Membership: |
Amplicon length greater than or equal to 101 Protein Kinase Detect Genomic DNA Rattus norvegicus Probe spans exons |