Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chia | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_207586.2 | NP_997469.1 | 5 - 6 | 505 | 61 | |
GenBank mRNA | AY486074.1 | - | 5 - 6 | 507 | 61 | |
KC529649.1 | - | 4 - 5 | 296 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |