Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Izumo4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006241036.2 | XP_006241098.1 | 3 - 4 | 785 | 58 | |
XM_008765151.1 | XP_008763373.1 | 3 - 4 | 785 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Rattus norvegicus Probe spans exons |