Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127635.2 | NP_001121107.1 | 1 - 2 | 43 | 76 | |
GenBank mRNA | BC166539.1 | - | 1 - 2 | 43 | 7191896 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |