Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gabarapl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044294.1 | NP_001037759.1 | 2 - 3 | 340 | 92 | |
GenBank mRNA | BC105627.1 | - | 2 - 3 | 340 | 7323087 | 92 |
FQ213425.1 | - | 2 - 3 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |