Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klrc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF021350.1 | - | 6 - 7 | 722 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Rattus norvegicus Probe spans exons |