Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spocd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008764193.1 | XP_008762415.1 | 4 - 5 | 370 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |