Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tcea3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015008.1 | NP_001015008.1 | 4 - 5 | 466 | 71 | |
GenBank mRNA | BC091180.1 | - | 4 - 5 | 466 | 7325833 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |