Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhdds | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011978.1 | NP_001011978.1 | 6 - 7 | 674 | 88 | |
XM_006239014.2 | XP_006239076.1 | 6 - 7 | 789 | 88 | ||
GenBank mRNA | BC085875.1 | - | 6 - 7 | 674 | 7191470 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |