Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pithd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_216543.7 | XP_216543.5 | 2 - 3 | 376 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |