Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pithd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_216543.7 | XP_216543.5 | 4 - 5 | 545 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |