Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem38b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014191.1 | NP_001014213.1 | 2 - 3 | 286 | 83 | |
GenBank mRNA | BC079336.1 | - | 2 - 3 | 286 | 7106021 | 83 |
FQ229075.1 | - | 2 - 3 | 327 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |