Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106651.1 | NP_001100121.1 | 2 - 3 | 568 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |