Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stk4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107800.1 | NP_001101270.1 | 1 - 2 | 91 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 Apoptosis Protein Kinase Rattus norvegicus Probe spans exons |