Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfatc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107805.1 | NP_001101275.1 | 2 - 3 | 1255 | 51 | |
XM_006235653.1 | XP_006235715.1 | 2 - 3 | 1249 | 51 | ||
XM_008762507.1 | XP_008760729.1 | 2 - 3 | 1249 | 51 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |