Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105813.1 | NP_001099283.1 | 17 - 18 | 1998 | 53 | |
XM_006246899.2 | XP_006246961.1 | 16 - 17 | 1917 | 53 | ||
GenBank mRNA | BC168148.1 | - | 16 - 17 | 1913 | 7443082 | 53 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus Probe spans exons |