Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kctd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105768.1 | NP_001099238.1 | 2 - 3 | 382 | 135 | |
GenBank mRNA | BC168712.1 | - | 2 - 3 | 385 | 8366833 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 Ion Channel Mammalian Gene Collection Rattus norvegicus Probe spans exons |